missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidocoumarin 98.0+%, TCI America™

Catalog No. p-7118174
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2972200MG 200 mg
A29721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2972200MG Supplier TCI America Supplier No. A2972200MG
Only null left

Chemical Identifiers

CAS 779-30-6
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.20
MDL Number MFCD00075556
InChI Key XJYLCCJIDYSFMB-UHFFFAOYSA-N
PubChem CID 136620
IUPAC Name N-(2-oxo-2H-chromen-3-yl)acetamide
SMILES CC(=O)NC1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 208°C
Quantity 200 mg
Formula Weight 203.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetamidocoumarin