missing translation for 'onlineSavingsMsg'
Learn More

3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 98.0+%, TCI America™

Catalog No. D102725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D102725G 25 g
D1027100G 100 g
D1027500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D102725G Supplier TCI America Supplier No. D102725G

Chemical Identifiers

CAS 78-19-3
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.25
MDL Number MFCD00006564
InChI Key OOXMQACSWCZQLX-UHFFFAOYSA-N
Synonym 3,9-divinyl-2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-divinylspirobi m-dioxane, diallylidene pentaerythritol, diallylidenepentaerythritol, pentaerythritol diacrolein acetal, acrolein pentaerythritol bisacetal, acrolein, cyclic diacetal with pentaerythritol, acrolein-pentaerythritol dicyclic acetal, acrolein, cyclic neopentanetetrayl acetal, 2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-diethenyl
PubChem CID 66218
IUPAC Name 3,9-diethenyl-2,4,8,10-tetraoxaspiro[5.5]undecane
SMILES C=CC1OCC2(CO1)COC(OC2)C=C

Specifications

Melting Point 44°C
Color White
Boiling Point 94°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane
EINECSNumber (5)-3770
RTECSNumber XF0875000
TSCA No