missing translation for 'onlineSavingsMsg'
Learn More

(+)-3,9-Dibromocamphor 97.0+%, TCI America™

Catalog No. D27155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27155G 5 g
D271525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27155G Supplier TCI America Supplier No. D27155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10293-10-4
Molecular Formula C10H14Br2O
Molecular Weight (g/mol) 310.029
MDL Number MFCD00167983
InChI Key DCDNKSJBRIJYEC-MZIHZIJQSA-N
PubChem CID 13783905
IUPAC Name (1S,2S,4R)-2-bromo-7-(bromomethyl)-4,7-dimethylbicyclo[2.2.1]heptan-3-one
SMILES CC12CCC(C1(C)CBr)C(C2=O)Br

Specifications

Color White
Quantity 5 g
Formula Weight 310.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-3,9-Dibromocamphor