missing translation for 'onlineSavingsMsg'
Learn More

3,9-Bis[2-(3,5-diamino-2,4,6-triazaphenyl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecane 98.0+%, TCI America™

Catalog No. B172425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B172425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B172425G Supplier TCI America Supplier No. B172425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22535-90-6
Molecular Formula C17H26N10O4
Molecular Weight (g/mol) 434.461
MDL Number MFCD00191395
InChI Key DUZLHGMYNVZMCO-UHFFFAOYSA-N
Synonym CTU Guanamine
PubChem CID 89744
IUPAC Name 6-[2-[3-[2-(4,6-diamino-1,3,5-triazin-2-yl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]ethyl]-1,3,5-triazine-2,4-diamine
SMILES C1C2(COC(O1)CCC3=NC(=NC(=N3)N)N)COC(OC2)CCC4=NC(=NC(=N4)N)N

Specifications

Color White
Quantity 25 g
Formula Weight 434.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Bis[2-(3,5-diamino-2,4,6-triazaphenyl)ethyl]-2,4,8,10-tetraoxaspiro[5.5]undecane
EINECSNumber (5)-1034
TSCA No