missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,9'-Bicarbazole 98.0+%, TCI America™
$102.52 - $512.62
Chemical Identifiers
| CAS | 18628-07-4 |
|---|---|
| Molecular Formula | C24H16N2 |
| Molecular Weight (g/mol) | 332.406 |
| MDL Number | MFCD27644690 |
| InChI Key | FHJJVSJWFYYPAC-UHFFFAOYSA-N |
| PubChem CID | 13236022 |
| IUPAC Name | 3-carbazol-9-yl-9H-carbazole |
| SMILES | C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)N4C5=CC=CC=C5C6=CC=CC=C64 |
Chemical Identifiers
| 18628-07-4 | |
| 332.406 | |
| FHJJVSJWFYYPAC-UHFFFAOYSA-N | |
| 3-carbazol-9-yl-9H-carbazole |
| C24H16N2 | |
| MFCD27644690 | |
| 13236022 | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)N4C5=CC=CC=C5C6=CC=CC=C64 |
Specifications
| 18628-07-4 | |
| C24H16N2 | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)N4C5=CC=CC=C5C6=CC=CC=C64 | |
| 332.406 | |
| 332.41 | |
| Crystalline Powder |
| 216°C | |
| MFCD27644690 | |
| FHJJVSJWFYYPAC-UHFFFAOYSA-N | |
| 3-carbazol-9-yl-9H-carbazole | |
| 13236022 | |
| ≥98.0% (HPLC,N) | |
| 3,9′-Bicarbazole |
Safety and Handling
TSCA : No