missing translation for 'onlineSavingsMsg'
Learn More

3-[(6-Methylpyrazin-2-yl)oxy]aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62614DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC62614DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

915707-61-8
201.229
ZFDWDKWQAXTCSU-UHFFFAOYSA-N
24229682
CC1=CN=CC(=N1)OC2=CC=CC(=C2)N
C11H11N3O
MFCD09702406
3-6-methylpyrazin-2-yl oxy aniline, benzenamine, 3-6-methyl-2-pyrazinyl oxy, 3-6-methylpyrazin-2-yloxy phenylamine, benzenamine,3-6-methyl-2-pyrazinyl oxy
3-(6-methylpyrazin-2-yl)oxyaniline

Specifications

3-[(6-Methylpyrazin-2-yl)oxy]aniline
96.5
97%
C11H11N3O
MFCD09702406
ZFDWDKWQAXTCSU-UHFFFAOYSA-N
3-(6-methylpyrazin-2-yl)oxyaniline
24229682
97%
915707-61-8
100.0
Amber Glass Bottle
1g
3-6-methylpyrazin-2-yl oxy aniline, benzenamine, 3-6-methyl-2-pyrazinyl oxy, 3-6-methylpyrazin-2-yloxy phenylamine, benzenamine,3-6-methyl-2-pyrazinyl oxy
CC1=CN=CC(=N1)OC2=CC=CC(=C2)N
201.229
201.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents