missing translation for 'onlineSavingsMsg'
Learn More

3,6-Diphenyl-1,2,4,5-tetrazine 98.0+%, TCI America™

Catalog No. D31755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31755G 5 g
D31751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31755G Supplier TCI America Supplier No. D31755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6830-78-0
Molecular Formula C14H10N4
Molecular Weight (g/mol) 234.26
MDL Number MFCD00023646
InChI Key XAUWSIIGUUMHQQ-UHFFFAOYSA-N
Synonym diphenyl-s-tetrazine, 3,6-diphenyl-s-tetrazine, 3,6-diphenyltetrazine, 1,2,4,5-tetrazine, 3,6-diphenyl, s-tetrazine, 2,6-diphenyl, s-tetrazine, 3,6-diphenyl, 3,6-diphenyl-sym-tetrazine, diphenyl-1,2,4,5-tetrazine, acmc-1baef, enamine_004961
PubChem CID 23263
IUPAC Name diphenyl-1,2,4,5-tetrazine
SMILES C1=CC=C(C=C1)C1=NN=C(N=N1)C1=CC=CC=C1

Specifications

Melting Point 196°C
Quantity 5 g
Formula Weight 234.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diphenyl-1,2,4,5-tetrazine
RTECSNumber XF6863300
TSCA No