missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dioxa-1,8-octanedithiol 97.0+%, TCI America™

Catalog No. d264925g
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D264925G 25 g
D2649100G 100 g
D2649500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D264925G Supplier TCI America Supplier No. D264925G

Chemical Identifiers

CAS 14970-87-7
Molecular Formula C6H14O2S2
Molecular Weight (g/mol) 182.296
MDL Number MFCD00015873
InChI Key HCZMHWVFVZAHCR-UHFFFAOYSA-N
Synonym 1,2-Bis(2-mercaptoethoxy)ethane, 1,8-Dimercapto-3,6-dioxaoctane, Ethylene Glycol Bis(2-mercaptoethyl) Ether
PubChem CID 84733
IUPAC Name 2-[2-(2-sulfanylethoxy)ethoxy]ethanethiol
SMILES C(COCCS)OCCS

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
Formula Weight 182.30
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,6-Dioxa-1,8-octanedithiol
EINECSNumber (2)-2451
TSCA Yes