missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 3,6-Dimethyl-1,4-dioxane-2,5-dione
Description
3,6-Dimethyl-1,4-dioxane-2,5-dione
Specifications
Specifications
| CAS | 95-96-5 |
| Melting Point | 116°C to 119°C |
| Boiling Point | 142°C (8 mmHg) |
| Molecular Formula | C6H8O4 |
| Linear Formula | C6H8O4 |
| MDL Number | MFCD00011685 |
| Quantity | 100 g |
| Synonym | Lactide |
| Molecular Weight (g/mol) | 144.13 |
| Percent Purity | 99% |