missing translation for 'onlineSavingsMsg'
Learn More

4-Methyltetrahydropyran, stabilized with BHT

Catalog No. AC467320010
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467320010 1 L
AC467321000 100 mL
AC467320025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467320010 Supplier Thermo Scientific Chemicals Supplier No. 467320010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4717-96-8 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4717-96-8
Molecular Weight (g/mol) 100.16

Specifications

Density 0.8570 g/mL
Boiling Point 105.0°C
Flash Point 6.5°C
Molecular Formula C6H12O
Quantity 1 L
Specific Gravity 0.857
Solubility Information Solubility aq. soln.: 19.2 g/L (20°C).
Viscosity 0.78 mPa.s (20°C)
Formula Weight 100.16
Chemical Name or Material 4-Methyltetrahydropyran
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Highly flammable liquid and vapor.

Causes severe skin burns and eye damage.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 225-207-5

RUO – Research Use Only