missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%

Catalog No. AAH6445206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6445206 5 g
AAH6445203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6445206 Supplier Thermo Scientific Chemicals Supplier No. H6445206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 287944-16-5 | C11H19BO3 | 210.08 g/mol

It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.

Chemical Identifiers

CAS 287944-16-5
Molecular Formula C11H19BO3
Molecular Weight (g/mol) 210.08
MDL Number MFCD11052631
InChI Key DOSGEBYQRMBTGS-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
PubChem CID 11218053
IUPAC Name 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCOCC1

Specifications

Melting Point 63°C to 67°C
Color White
Quantity 5 g
Beilstein 8548328
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 210.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only