missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridine-2-carboxylic acid, 96%

Catalog No. AAH2714606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2714606 5 g
AAH2714603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2714606 Supplier Thermo Scientific Chemicals Supplier No. H2714606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1702-17-6 | C6H3Cl2NO2 | 192.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1702-17-6
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00078655
InChI Key HUBANNPOLNYSAD-UHFFFAOYSA-N
Synonym clopyralid, 3,6-dichloropicolinic acid, 3,6-dichloro-2-pyridinecarboxylic acid, lontrel, 2-pyridinecarboxylic acid, 3,6-dichloro, clopyralide, benzalox, cirtoxin, matrigon, transline
PubChem CID 15553
ChEBI CHEBI:62961
IUPAC Name 3,6-dichloropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(Cl)=CC=C1Cl

Specifications

Melting Point ∽150°C (decomposition)
Flash Point >100°C (212°F)
Quantity 5 g
Beilstein 473755
Merck Index 14,2400
Formula Weight 192
Percent Purity 96%
Chemical Name or Material 3,6-Dichloropyridine-2-carboxylic acid
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 216-935-4
RTECSNumber TJ7550700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only