missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridazine 98.0+%, TCI America™

Catalog No. D146525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D146525G 25 g
D1465500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D146525G Supplier TCI America Supplier No. D146525G

Chemical Identifiers

CAS 141-30-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006466
InChI Key GUSWJGOYDXFJSI-UHFFFAOYSA-N
Synonym pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
PubChem CID 67331
IUPAC Name 3,6-dichloropyridazine
SMILES C1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 69°C
Color White
Quantity 25 g
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dichloropyridazine
EINECSNumber (5)-3834
RTECSNumber UR5886250
TSCA Yes