missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloropyridazine, 97%

CAS: 141-30-0 | C4H2Cl2N2 | 148.98 g/mol

$212.49 - $212.49

Chemical Identifiers

CAS 141-30-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.98
MDL Number MFCD00006466
InChI Key GUSWJGOYDXFJSI-UHFFFAOYSA-N
Synonym pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
PubChem CID 67331
IUPAC Name 3,6-dichloropyridazine
SMILES C1=CC(=NN=C1Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC113711000
SDS View Documents
Thermo Scientific Chemicals
113711000
100 g Plastic bottle
Each for $212.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-30-0
148.98
GUSWJGOYDXFJSI-UHFFFAOYSA-N
67331
C1=CC(=NN=C1Cl)Cl
C4H2Cl2N2
MFCD00006466
pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
3,6-dichloropyridazine

Specifications

141-30-0
100.0
White
96% min. (GC)
C4H2Cl2N2
100 g
GUSWJGOYDXFJSI-UHFFFAOYSA-N
3,6-dichloropyridazine
67331
97%
3, 6-Dichloropyridazine
96.0
65°C to 69°C
Authentic
Plastic bottle
MFCD00006466
pyridazine, 3,6-dichloro, 3,6-dichloro-pyridazine, 3,6-dichloro-1,2-diazine, pubchem9487, 3,6-dichloropyridazin, 3,6 dichloropyridazine, 3.6-dichloropyridazine, pyridazine,6-dichloro, 3,6-dichloro-pyridazin, 3,6-dichloropyridazine
C1=CC(=NN=C1Cl)Cl
148.98
148.98
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear prote

GHS Signal Word: Danger

EINECSNumber : 205-478-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only