missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichlorocarbazole 96.0+%, TCI America™

Catalog No. p-7127892
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37511G 1 g
D37515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37511G Supplier TCI America Supplier No. D37511G

Chemical Identifiers

CAS 5599-71-3
Molecular Formula C12H7Cl2N
Molecular Weight (g/mol) 236.095
MDL Number MFCD00218271
InChI Key BIMDTNQPZWJKPL-UHFFFAOYSA-N
PubChem CID 236487
IUPAC Name 3,6-dichloro-9H-carbazole
SMILES C1=CC2=C(C=C1Cl)C3=C(N2)C=CC(=C3)Cl

Specifications

Melting Point 208°C
Quantity 1 g
Formula Weight 236.10
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dichlorocarbazole