missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-4-methylpyridazine 98.0+%, TCI America™

Catalog No. D42061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42061G 1 g
D42065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42061G Supplier TCI America Supplier No. D42061G

Chemical Identifiers

CAS 19064-64-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006465
InChI Key ROYHWGZNGMXQEU-UHFFFAOYSA-N
Synonym 4-methyl-3,6-dichloropyridazine, 3,6-dichloro-4-methyl-pyridazine, pyridazine, 3,6-dichloro-4-methyl, 3,6-dichloro-5-methylpyridazine, 3,6-dichloro-4-methyl-1,2-diazine, pubchem9497, ksc495e6l, 3,6-dichloro-4-methylpyridazin, pyridazine,6-dichloro-4-methyl, 3,6-di chloro-4-methyl pyridazine
PubChem CID 87923
IUPAC Name 3,6-dichloro-4-methylpyridazine
SMILES CC1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 151°C
Quantity 1 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dichloro-4-methylpyridazine