missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-4-methylpyridazine, 97%, Thermo Scientific Chemicals

Catalog No. p-7024784
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1999806 5 g
AAA1999814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1999806 Supplier Thermo Scientific Chemicals Supplier No. A1999806
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19064-64-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006465
InChI Key ROYHWGZNGMXQEU-UHFFFAOYSA-N
Synonym 4-methyl-3,6-dichloropyridazine, 3,6-dichloro-4-methyl-pyridazine, pyridazine, 3,6-dichloro-4-methyl, 3,6-dichloro-5-methylpyridazine, 3,6-dichloro-4-methyl-1,2-diazine, pubchem9497, ksc495e6l, 3,6-dichloro-4-methylpyridazin, pyridazine,6-dichloro-4-methyl, 3,6-di chloro-4-methyl pyridazine
PubChem CID 87923
IUPAC Name 3,6-dichloro-4-methylpyridazine
SMILES CC1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 86°C to 88°C
Boiling Point 149°C to 151°C (21 mmHg)
Quantity 5 g
Beilstein 119573
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 3,6-Dichloro-4-methylpyridazine
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-794-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only