missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. D41171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41171G Supplier TCI America Supplier No. D41171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity laboratory reagent for chemical research applications.

Chemical Identifiers

CAS 1702-17-6
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00078655
InChI Key HUBANNPOLNYSAD-UHFFFAOYSA-N
Synonym clopyralid, 3,6-dichloropicolinic acid, 3,6-dichloro-2-pyridinecarboxylic acid, lontrel, 2-pyridinecarboxylic acid, 3,6-dichloro, clopyralide, benzalox, cirtoxin, matrigon, transline
PubChem CID 15553
ChEBI CHEBI:62961
IUPAC Name 3,6-dichloropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(Cl)=CC=C1Cl

Specifications

Melting Point 151°C
Quantity 1 g
Formula Weight 192.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dichloro-2-pyridinecarboxylic Acid
RTECSNumber TJ7550700
TSCA Yes