missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(p-tolyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7128599
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5001200MG 200 mg
D50011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5001200MG Supplier TCI America Supplier No. D5001200MG
Only null left

Chemical Identifiers

CAS 357437-74-2
Molecular Formula C19H13Br2N
Molecular Weight (g/mol) 415.128
MDL Number MFCD12024282
InChI Key ZLCMXHALWTYHOY-UHFFFAOYSA-N
PubChem CID 53401077
IUPAC Name 3,6-dibromo-9-(4-methylphenyl)carbazole
SMILES CC1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 200 mg
Formula Weight 415.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(p-tolyl)-9H-carbazole