missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromo-9-n-octylcarbazole 98.0+%, TCI America™
$96.26 - $331.13
Chemical Identifiers
| CAS | 79554-93-1 |
|---|---|
| Molecular Formula | C20H23Br2N |
| Molecular Weight (g/mol) | 437.219 |
| MDL Number | MFCD23703112 |
| InChI Key | MFYGWCVLNPQWRR-UHFFFAOYSA-N |
| PubChem CID | 11339570 |
| IUPAC Name | 3,6-dibromo-9-octylcarbazole |
| SMILES | CCCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4482200MG
|
TCI America
D4482200MG |
200 mg |
Each for $96.26
|
|
|||||
|
D44821G
|
TCI America
D44821G |
1 g |
Each for $331.13
|
|
|||||
Chemical Identifiers
| 79554-93-1 | |
| 437.219 | |
| MFYGWCVLNPQWRR-UHFFFAOYSA-N | |
| 3,6-dibromo-9-octylcarbazole |
| C20H23Br2N | |
| MFCD23703112 | |
| 11339570 | |
| CCCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br |
Specifications
| 79554-93-1 | |
| White-Yellow | |
| MFCD23703112 | |
| MFYGWCVLNPQWRR-UHFFFAOYSA-N | |
| 3,6-dibromo-9-octylcarbazole | |
| 11339570 | |
| ≥98.0% (GC) | |
| 3,6-Dibromo-9-n-octylcarbazole |
| 83°C | |
| C20H23Br2N | |
| 200 mg | |
| CCCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br | |
| 437.219 | |
| 437.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No