missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-n-octylcarbazole 98.0+%, TCI America™

Catalog No. D4482200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4482200MG 200 mg
D44821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4482200MG Supplier TCI America Supplier No. D4482200MG
Only null left

Chemical Identifiers

CAS 79554-93-1
Molecular Formula C20H23Br2N
Molecular Weight (g/mol) 437.219
MDL Number MFCD23703112
InChI Key MFYGWCVLNPQWRR-UHFFFAOYSA-N
PubChem CID 11339570
IUPAC Name 3,6-dibromo-9-octylcarbazole
SMILES CCCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 200 mg
Formula Weight 437.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-n-octylcarbazole