missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-heptyl-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7128759
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54041G 1 g
D54045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54041G Supplier TCI America Supplier No. D54041G
Only null left

Chemical Identifiers

CAS 1103535-99-4
Molecular Formula C19H21Br2N
Molecular Weight (g/mol) 423.192
InChI Key DTRIORAMBSBROF-UHFFFAOYSA-N
PubChem CID 129319316
IUPAC Name 3,6-dibromo-9-heptylcarbazole
SMILES CCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 423.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-heptyl-9H-carbazole