missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-ethylcarbazole 98.0+%, TCI America™

Catalog No. D29821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29821G 1 g
D29825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29821G Supplier TCI America Supplier No. D29821G
Only null left

Chemical Identifiers

CAS 33255-13-9
Molecular Formula C14H11Br2N
Molecular Weight (g/mol) 353.057
InChI Key GZBJRMVGNVDUCO-UHFFFAOYSA-N
PubChem CID 631855
IUPAC Name 3,6-dibromo-9-ethylcarbazole
SMILES CCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br

Specifications

Melting Point 141°C
Quantity 1 g
Formula Weight 353.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-ethylcarbazole