missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. D5341200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5341200MG 200 mg
D53411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5341200MG Supplier TCI America Supplier No. D5341200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 917773-26-3
Molecular Formula C26H27Br2NO
Molecular Weight (g/mol) 529.32
MDL Number MFCD30536723
InChI Key PHKYODAUSDAEKX-UHFFFAOYSA-N
PubChem CID 71428395
IUPAC Name 3,6-dibromo-9-[4-(octyloxy)phenyl]-9H-carbazole
SMILES CCCCCCCCOC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 78°C
Quantity 200 mg
Formula Weight 529.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole