missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. D53401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53401G 1 g
D53405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53401G Supplier TCI America Supplier No. D53401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 746651-52-5
Molecular Formula C19H13Br2NO
Molecular Weight (g/mol) 431.127
InChI Key YXWHLCSWBLHWCH-UHFFFAOYSA-N
PubChem CID 58620860
IUPAC Name 3,6-dibromo-9-(4-methoxyphenyl)carbazole
SMILES COC1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 1 g
Formula Weight 431.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole