missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole 95.0+%, TCI America™
Supplier: TCI America D5342200MG
Specifications
| 3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole | |
| 59°C | |
| MFCD30721573 | |
| ACUVALSRGHGNLZ-UHFFFAOYNA-N | |
| 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole | |
| 68106047 | |
| ≥95.0% (HPLC,N) |
| 946491-48-1 | |
| C26H27Br2NO | |
| 200 mg | |
| CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 | |
| 529.32 | |
| 529.32 | |
| Crystalline Powder |
Chemical Identifiers
| 946491-48-1 | |
| 529.32 | |
| ACUVALSRGHGNLZ-UHFFFAOYNA-N | |
| 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole |
| C26H27Br2NO | |
| MFCD30721573 | |
| 68106047 | |
| CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 |
Safety and Handling
TSCA : No