missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole 97.0+%, TCI America™

Catalog No. p-7128764
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5421200MG 200 mg
D54211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5421200MG Supplier TCI America Supplier No. D5421200MG
Only null left

Chemical Identifiers

CAS 1873364-08-9
Molecular Formula C20H15Br2N
Molecular Weight (g/mol) 429.16
MDL Number MFCD31618134
InChI Key ASACIWHKQZICTN-UHFFFAOYSA-N
PubChem CID 129319319
IUPAC Name 3,6-dibromo-9-(3,5-dimethylphenyl)-9H-carbazole
SMILES CC1=CC(=CC(C)=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 240°C
Quantity 200 mg
Formula Weight 429.16
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole