missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(2-ethylhexyl)carbazole 98.0+%, TCI America™

Catalog No. D48305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48305G 5 g
D48301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48305G Supplier TCI America Supplier No. D48305G
Only null left

Chemical Identifiers

CAS 173063-52-0
Molecular Formula C20H23Br2N
Molecular Weight (g/mol) 437.22
MDL Number MFCD18252907
InChI Key ZDFZWZGIGKUBRA-UHFFFAOYNA-N
PubChem CID 16752909
IUPAC Name 3,6-dibromo-9-(2-ethylhexyl)-9H-carbazole
SMILES CCCCC(CC)CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 437.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,6-Dibromo-9-(2-ethylhexyl)carbazole