missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-2-chloropyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH2773803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2773803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2773803 Supplier Thermo Scientific Chemicals Supplier No. H2773803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 942206-18-0
Molecular Formula C5H2Br2ClN
Molecular Weight (g/mol) 271.336
MDL Number MFCD09037464
InChI Key CVZSNXGMWIGPBF-UHFFFAOYSA-N
Synonym 2-chloro-3,6-dibromopyridine, 3,6-dibromo-2-chloro-pyridine
PubChem CID 42553063
IUPAC Name 3,6-dibromo-2-chloropyridine
SMILES C1=CC(=NC(=C1Br)Cl)Br

Specifications

Melting Point 84°C to 86°C
Quantity 1 g
Formula Weight 271.34
Percent Purity 95%
Chemical Name or Material 3,6-Dibromo-2-chloropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only