missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di(4-pyridyl)-1,2,4,5-tetrazine 98.0+%, TCI America™

Catalog No. D32111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32111G 1 g
D32115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32111G Supplier TCI America Supplier No. D32111G
Only null left

Chemical Identifiers

CAS 57654-36-1
Molecular Formula C12H8N6
Molecular Weight (g/mol) 236.24
MDL Number MFCD00129054
InChI Key MQSMIIJCRWDMDL-UHFFFAOYSA-N
Synonym 3,6-Bis(4-pyridyl)-1,2,4,5-tetrazine
PubChem CID 4980025
IUPAC Name bis(pyridin-4-yl)-1,2,4,5-tetrazine
SMILES C1=CC(=CC=N1)C1=NN=C(N=N1)C1=CC=NC=C1

Specifications

Melting Point 257°C
Color Red-Yellow
Quantity 1 g
Formula Weight 236.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Di(4-pyridyl)-1,2,4,5-tetrazine