missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95.0+%, TCI America™

Catalog No. d39695g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39695G 5 g
D39691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39695G Supplier TCI America Supplier No. D39695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 850583-75-4
Molecular Formula C14H8N2O2S2
Molecular Weight (g/mol) 300.35
InChI Key YIUHGBNJJRTMIE-UHFFFAOYSA-N
PubChem CID 53384408
IUPAC Name 1,4-dithiophen-2-yl-2,5-dihydropyrrolo[3,4-c]pyrrole-3,6-dione
SMILES C1=CSC(=C1)C2=C3C(=C(NC3=O)C4=CC=CS4)C(=O)N2

Specifications

Color Black
Quantity 5 g
Formula Weight 300.35
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione