missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95.0+%, TCI America™
Specifications
Specifications
| Color | Black |
| Quantity | 1 g |
| Formula Weight | 300.35 |
| Percent Purity | ≥95.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione |
Safety and Handling
| TSCA | No |