missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 98.0+%, TCI America™

Catalog No. D36401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36401G 1 g
D36405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36401G Supplier TCI America Supplier No. D36401G

Chemical Identifiers

CAS 1671-87-0
Molecular Formula C12H8N6
Molecular Weight (g/mol) 236.24
MDL Number MFCD00121717
InChI Key JFBIRMIEJBPDTQ-UHFFFAOYSA-N
Synonym 3,6-di pyridin-2-yl-1,2,4,5-tetrazine, 3,6-di 2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridyl-1,2,4,5-tetrazine, 3,6-di-2-pyridinyl-1,2,4,5-tetrazine, 3,6-bis 2-pyridyl-1,2,4,5-tetrazine, bis pyridin-2-yl-1,2,4,5-tetrazine, 3,6-bis 2-pyridinyl-1,2,4,5-tetrazine, bptz, acmc-209dvt, 3,6-di-2-pyridyl-s-tetrazine
PubChem CID 315109
IUPAC Name bis(pyridin-2-yl)-1,2,4,5-tetrazine
SMILES C1=CC=C(N=C1)C1=NN=C(N=N1)C1=CC=CC=N1

Specifications

Melting Point 225°C
Color Red
Quantity 1 g
Formula Weight 236.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Di(2-pyridyl)-1,2,4,5-tetrazine