missing translation for 'onlineSavingsMsg'
Learn More

[3-(6-Aminohexylamino)propyl]trimethoxysilane 97.0+%, TCI America™

Catalog No. a31285ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31285ML 5 mL
A312825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31285ML Supplier TCI America Supplier No. A31285ML
Only null left

Chemical Identifiers

CAS 51895-58-0
Molecular Formula C12H30N2O3Si
Molecular Weight (g/mol) 278.47
MDL Number MFCD00054943
InChI Key AMVXVPUHCLLJRE-UHFFFAOYSA-N
Synonym N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine, N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine
PubChem CID 9925638
IUPAC Name (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine
SMILES CO[Si](CCCNCCCCCCN)(OC)OC

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 mL
Formula Weight 278.47
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [3-(6-Aminohexylamino)propyl]trimethoxysilane