missing translation for 'onlineSavingsMsg'
Learn More

3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB05684EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB05684EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

175135-15-6
244.217
FDRMNTVCDKSRNL-UHFFFAOYSA-N
2775098
C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO
C11H11F3N2O
MFCD00067735
3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol

Specifications

3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol
96.5
97%
C11H11F3N2O
MFCD00067735
FDRMNTVCDKSRNL-UHFFFAOYSA-N
3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol
2775098
97%
175135-15-6
100.0
Amber Glass Bottle
10g
3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO
244.217
244.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents