missing translation for 'onlineSavingsMsg'
Learn More

3,5-Pyridinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134670
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05515G 5 g
P055125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P05515G Supplier TCI America Supplier No. P05515G
Only null left

Chemical Identifiers

CAS 499-81-0
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006393
InChI Key MPFLRYZEEAQMLQ-UHFFFAOYSA-N
Synonym 3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f
PubChem CID 10366
ChEBI CHEBI:46875
IUPAC Name pyridine-3,5-dicarboxylic acid
SMILES OC(=O)C1=CC(=CN=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Pyridinedicarboxylic Acid