missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-2-furyl)propionaldehyde 98.0+%, TCI America™

Catalog No. M24985G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24985G Supplier TCI America Supplier No. M24985G
Only null left

Chemical Identifiers

CAS 34756-16-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD01910105
InChI Key WPBUABKBDHGOAJ-UHFFFAOYSA-N
Synonym 3-(5-Methyl-2-furyl)propanal
PubChem CID 99537
IUPAC Name 3-(5-methylfuran-2-yl)propanal
SMILES CC1=CC=C(O1)CCC=O

Specifications

Color Yellow
Boiling Point 58°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(5-Methyl-2-furyl)propionaldehyde
EINECSNumber (5)-3345
TSCA No