missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline, 97%, Thermo Scientific™

$190.67 - $190.67

Chemical Identifiers

CAS 10185-69-0
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD08443707
InChI Key CTRGRIHPFAVSOF-UHFFFAOYSA-N
Synonym 3-5-methyl-1,2,4-oxadiazol-3-yl aniline, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylamine, benzenamine,3-5-methyl-1,2,4-oxadiazol-3-yl
PubChem CID 11423869
IUPAC Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
SMILES CC1=NC(=NO1)C1=CC(N)=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC39114DA
SDS View Documents
Thermo Scientific Chemicals
CC39114DA
1 g
Each for $190.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10185-69-0
175.19
CTRGRIHPFAVSOF-UHFFFAOYSA-N
11423869
CC1=NC(=NO1)C1=CC(N)=CC=C1
C9H9N3O
MFCD08443707
3-5-methyl-1,2,4-oxadiazol-3-yl aniline, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylamine, benzenamine,3-5-methyl-1,2,4-oxadiazol-3-yl
3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline

Specifications

10185-69-0
100.0
Amber glass bottle
MFCD08443707
3-5-methyl-1,2,4-oxadiazol-3-yl aniline, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylamine, benzenamine,3-5-methyl-1,2,4-oxadiazol-3-yl
CC1=NC(=NO1)C1=CC(N)=CC=C1
175.19
175.19
3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)aniline
96.5
97%
C9H9N3O
1 g
CTRGRIHPFAVSOF-UHFFFAOYSA-N
3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
11423869
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents