missing translation for 'onlineSavingsMsg'
Learn More

3,5-Lutidine 98.0+%, TCI America™

Catalog No. L006825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L006825ML 25 mL
L0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L006825ML Supplier TCI America Supplier No. L006825ML

Chemical Identifiers

CAS 591-22-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006404
InChI Key HWWYDZCSSYKIAD-UHFFFAOYSA-N
Synonym 3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825
PubChem CID 11565
IUPAC Name 3,5-dimethylpyridine
SMILES CC1=CC(=CN=C1)C

Specifications

Melting Point -9°C
Color Yellow
Boiling Point 172°C
Quantity 25 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Lutidine
EINECSNumber (5)-0712
TSCA Yes