Learn More
3,5-Lutidine, 99%
CAS: 591-22-0 | C7H9N | 107.156 g/mol
Supplier: Thermo Scientific Chemicals A14877AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,5-Lutidine | |
| -9°C | |
| 169°C to 170°C | |
| Unpleasant | |
| 1.505 | |
| 100 mL | |
| 105682 | |
| Sol | |
| CC1=CC(=CN=C1)C | |
| 107.156 | |
| 107.16 |
| 591-22-0 | |
| 0.941 | |
| 53°C (127°F) | |
| C7H9N | |
| MFCD00006404 | |
| UN1992 | |
| 3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825 | |
| HWWYDZCSSYKIAD-UHFFFAOYSA-N | |
| 3,5-dimethylpyridine | |
| 11565 | |
| 99% |
Chemical Identifiers
| 591-22-0 | |
| 107.156 | |
| HWWYDZCSSYKIAD-UHFFFAOYSA-N | |
| 11565 | |
| CC1=CC(=CN=C1)C |
| C7H9N | |
| MFCD00006404 | |
| 3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825 | |
| 3,5-dimethylpyridine |
Safety and Handling
GHS H Statement
H301-H331-H226-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H331-H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 209-708-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only