missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diphenylpyrazole 98.0+%, TCI America™
$128.62 - $508.89
Chemical Identifiers
| CAS | 1145-01-3 |
|---|---|
| Molecular Formula | C15H12N2 |
| Molecular Weight (g/mol) | 220.28 |
| MDL Number | MFCD00039675 |
| InChI Key | JXHKUYQCEJILEI-UHFFFAOYSA-N |
| Synonym | 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g |
| PubChem CID | 70840 |
| IUPAC Name | 3,5-diphenyl-1H-pyrazole |
| SMILES | N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1145-01-3 | |
| 220.28 | |
| JXHKUYQCEJILEI-UHFFFAOYSA-N | |
| 70840 | |
| N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H12N2 | |
| MFCD00039675 | |
| 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g | |
| 3,5-diphenyl-1H-pyrazole |
Specifications
| 1145-01-3 | |
| Yellow | |
| MFCD00039675 | |
| 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g | |
| N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
| 220.28 | |
| 220.28 | |
| Crystalline Powder |
| 202°C | |
| C15H12N2 | |
| 5 g | |
| JXHKUYQCEJILEI-UHFFFAOYSA-N | |
| 3,5-diphenyl-1H-pyrazole | |
| 70840 | |
| ≥98.0% (GC) | |
| 3,5-Diphenylpyrazole |
Safety and Handling
TSCA : Yes