missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diphenylpyrazole, 99%
CAS: 1145-01-3 | C15H12N2 | 220.28 g/mol
$114.55 - $114.55
Chemical Identifiers
| CAS | 1145-01-3 |
|---|---|
| Molecular Formula | C15H12N2 |
| Molecular Weight (g/mol) | 220.28 |
| MDL Number | MFCD00039675 |
| InChI Key | JXHKUYQCEJILEI-UHFFFAOYSA-N |
| Synonym | 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g |
| PubChem CID | 70840 |
| IUPAC Name | 3,5-diphenyl-1H-pyrazole |
| SMILES | N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC337870050
|
Thermo Scientific Chemicals
337870050 |
5 g | Glass bottle |
Each for $114.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1145-01-3 | |
| 220.28 | |
| JXHKUYQCEJILEI-UHFFFAOYSA-N | |
| 70840 | |
| N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H12N2 | |
| MFCD00039675 | |
| 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g | |
| 3,5-diphenyl-1H-pyrazole |
Specifications
| 1145-01-3 | |
| 100.0 | |
| White | |
| 98.5% min. (HPLC) | |
| C15H12N2 | |
| 5 g | |
| JXHKUYQCEJILEI-UHFFFAOYSA-N | |
| 3,5-diphenyl-1H-pyrazole | |
| 70840 | |
| 99% | |
| 3, 5-Diphenylpyrazole, 99% |
| 98.5 | |
| 200.0°C to 203.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00039675 | |
| 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g | |
| N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
| 220.28 | |
| 220.27 | |
| Crystalline Powder |
Safety and Handling
LIGHT SENSITIVE
EINECSNumber : 214-543-8
TSCA : TSCA
RUO – Research Use Only