missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diphenylpyrazole, 99%

Catalog No. AC337870050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337870050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC337870050 Supplier Thermo Scientific Chemicals Supplier No. 337870050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1145-01-3 | C15H12N2 | 220.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1145-01-3
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00039675
InChI Key JXHKUYQCEJILEI-UHFFFAOYSA-N
Synonym 3,5-diphenylpyrazole, 1h-pyrazole, 3,5-diphenyl, pyrazole, 3,5-diphenyl, pyrazole,5-diphenyl, acmc-1bufo, 1h-pyrazole,5-diphenyl, cbmicro_048085, 3,5-diphenyl pyrazole, 3,5-diphenyl-1h-pyrazole 5g
PubChem CID 70840
IUPAC Name 3,5-diphenyl-1H-pyrazole
SMILES N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 200.0°C to 203.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 220.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Diphenylpyrazole, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 214-543-8
TSCA TSCA

RUO – Research Use Only