missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrrole-2-carboxaldehyde, 97%

Catalog No. AAH50478MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50478MD 250 mg
AAH5047803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50478MD Supplier Thermo Scientific Chemicals Supplier No. H50478MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2199-58-8 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2199-58-8
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00111522
InChI Key RDFZYUOHJBXMJA-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole
PubChem CID 270465
IUPAC Name 3,5-dimethyl-1H-pyrrole-2-carbaldehyde
SMILES CC1=CC(=C(N1)C=O)C

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylpyrrole-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only