missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyridine-4-boronic acid pinacol ester, 95%

Catalog No. AAH32159ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32159ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32159ME Supplier Thermo Scientific Chemicals Supplier No. H32159ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1032358-02-3 | C13H20BNO2 | 233.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1032358-02-3
Molecular Formula C13H20BNO2
Molecular Weight (g/mol) 233.12
MDL Number MFCD10566506
InChI Key PLCYEDJFSPVVLL-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3,5-dimethylpyridine-4-boronic acid pinacol ester, 2-3,5-dimethyl-4-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 45588136
IUPAC Name 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1=CN=CC(C)=C1B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 500 mg
Formula Weight 233.12
Percent Purity 95%
Chemical Name or Material 3,5-Dimethylpyridine-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only