missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazin-2-amine, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB07911EA

Quantity 10g

 View more versions of this product

Catalog No. BTB07911EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

91678-81-8
123.159
GZZRWBRYYZGTNW-UHFFFAOYSA-N
2774764
CC1=CN=C(C(=N1)C)N
C6H9N3
MFCD00107172
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
3,5-dimethylpyrazin-2-amine

Specifications

3, 5-Dimethylpyrazin-2-amine
96.5
97+%
10g
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
CC1=CN=C(C(=N1)C)N
123.159
123.16
91678-81-8
100.0
C6H9N3
MFCD00107172
GZZRWBRYYZGTNW-UHFFFAOYSA-N
3,5-dimethylpyrazin-2-amine
2774764
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents