missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazin-2-amine, ≥97%, Thermo Scientific™

Catalog No. BTB07911DA
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB07911DA 1g
BTB07911EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB07911DA Supplier Thermo Scientific Chemicals Supplier No. BTB07911DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91678-81-8
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00107172
InChI Key GZZRWBRYYZGTNW-UHFFFAOYSA-N
Synonym 2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
PubChem CID 2774764
IUPAC Name 3,5-dimethylpyrazin-2-amine
SMILES CC1=CN=C(C(=N1)C)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethylpyrazin-2-amine
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.