missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpiperidine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7126719
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141125ML 25 mL
D1411500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D141125ML Supplier TCI America Supplier No. D141125ML
Only null left

Chemical Identifiers

CAS 35794-11-7
Molecular Formula C7H16N
Molecular Weight (g/mol) 114.21
MDL Number MFCD00005996,MFCD09832871
InChI Key IDWRJRPUIXRFRX-KNVOCYPGSA-O
Synonym piperidine, 3,5-dimethyl, 3,5-lupetidine, 3,5-dimethylpiperidin, 3,5-dimethylpiperidine, cis + trans, pubchem7709, 3,5-dimethylpiperdine, 3,5 dimethylpiperidine, 3,5-dimethylpiperadine, acmc-1afvq, hexahydro-3,5-lutidine
PubChem CID 118259
IUPAC Name (3R,5S)-3,5-dimethylpiperidin-1-ium
SMILES C[C@H]1C[NH2+]C[C@@H](C)C1

Specifications

Color Colorless
Boiling Point 144°C
Quantity 25 mL
UN Number 1993
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylpiperidine (cis- and trans- mixture)
EINECSNumber (5)-0769
TSCA Yes