missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole 99.0+%, TCI America™

Catalog No. p-7126919
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176725ML 25 mL
D1767250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D176725ML Supplier TCI America Supplier No. D176725ML
Only null left

Chemical Identifiers

CAS 300-87-8
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003156
InChI Key FICAQKBMCKEFDI-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole, isoxazole, 3,5-dimethyl, 3,5-dimethylisooxazole, 3,5-dwumetyloizoksazolu, 3,5-dwumetyloizoksazolu polish, 3,5-dimethylisoxazol, pubchem8641, isoxazole,5-dimethyl, 3,5-dimethyl-isoxazole
PubChem CID 9312
IUPAC Name 3,5-dimethyl-1,2-oxazole
SMILES CC1=CC(=NO1)C

Specifications

Color Yellow
Boiling Point 141°C
Quantity 25 mL
UN Number 1993
Formula Weight 97.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylisoxazole
RTECSNumber NY2774200
TSCA Yes