missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylisoxazole, 99%
Description
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | -14°C |
| Density | 0.992 |
| Boiling Point | 142°C to 144°C |
| Flash Point | 31°C (87°F) |
| Refractive Index | 1.4415 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 106324 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 97.12 |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 206-100-2 |
| RTECSNumber | NY2774200 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only